Learn More
MSC SPI Radius Gage, 7/16 Inch, Satin Chrome Finish, Steel
Catalog No. 5000512371
Encompass
The SPI 7/16 Inch Radius Gage is designed for precise measurement of convex and concave radii. Constructed from durable steel with a satin chrome finish, it provides reliable performance in various applications.
  • Features a 7/16 inch radius for accurate measurements.
  • Equipped with a satin chrome finish for durability and readability.
  • Made of steel, ensuring a long lifespan.
  • Includes 5 measuring surfaces for versatile use.
Catalog No. 50-005-12371 Supplier MSC Supplier No. 01718535
The SPI 7/16 Inch Radius Gage is designed for precise measurement of convex and concave radii. Constructed from durable steel with a satin chrome finish, it provides reliable performance in various applications.
  • Features a 7/16 inch radius for accurate measurements.
  • Equipped with a satin chrome finish for durability and readability.
  • Made of steel, ensuring a long lifespan.
  • Includes 5 measuring surfaces for versatile use.