Learn More
MSC SPI Radius Gage Set, Stainless Steel, 0.01-0.5 in, 26 pc
Catalog No. 5000529118
Encompass
This radius gage set is made of stainless steel and includes 26 pieces, designed to measure radii from 0.01 to 0.5 inches. It features a satin chrome finish and comes with a protective vinyl case and holder, making it suitable for precise radius measurement in various applications.
  • Constructed from stainless steel for durability.
  • Measures radii ranging from 0.01 to 0.5 inches.
  • Features a satin chrome finish for enhanced visibility and protection.
  • Includes a holder for organization.
  • Comes with a protective vinyl case for storage and transport.
  • Offers an accuracy of +/-0.0012 "
  • Contains 26 individual gages for comprehensive measurement.
Catalog No. 50-005-29118 Supplier MSC Supplier No. 01718766
This radius gage set is made of stainless steel and includes 26 pieces, designed to measure radii from 0.01 to 0.5 inches. It features a satin chrome finish and comes with a protective vinyl case and holder, making it suitable for precise radius measurement in various applications.
  • Constructed from stainless steel for durability.
  • Measures radii ranging from 0.01 to 0.5 inches.
  • Features a satin chrome finish for enhanced visibility and protection.
  • Includes a holder for organization.
  • Comes with a protective vinyl case for storage and transport.
  • Offers an accuracy of +/-0.0012 "
  • Contains 26 individual gages for comprehensive measurement.