Learn More
MSC SPI Radius Gage Set, Stainless Steel, 9/16 to 1 in Radius, 8 pc, Satin Chrome Finish
Catalog No. 5000529843
Encompass
This radius gage set is a precision measuring tool crafted from durable stainless steel. This 8-piece set is designed to accurately measure radii from 9/16 to 1 in, with an impressive accuracy of +/-0.0012 decimal inches. Made in Japan, this radius gage set ensures reliable performance for various measurement applications.
  • Crafted from durable stainless steel
  • 8-piece set accurately measures radii from 9/16 to 1 in
  • Accuracy of +/-0.0012 decimal inches
  • Includes gages for radii of 7/8, 1, 9/16, 13/16, 3/4, 15/16, 11/16, and 5/8 in
  • Each gage features a satin chrome finish for enhanced durability and readability
  • Comes complete with a protective vinyl case for convenient storage and transport
Catalog No. 50-005-29843 Supplier MSC Supplier No. 01718469
This radius gage set is a precision measuring tool crafted from durable stainless steel. This 8-piece set is designed to accurately measure radii from 9/16 to 1 in, with an impressive accuracy of +/-0.0012 decimal inches. Made in Japan, this radius gage set ensures reliable performance for various measurement applications.
  • Crafted from durable stainless steel
  • 8-piece set accurately measures radii from 9/16 to 1 in
  • Accuracy of +/-0.0012 decimal inches
  • Includes gages for radii of 7/8, 1, 9/16, 13/16, 3/4, 15/16, 11/16, and 5/8 in
  • Each gage features a satin chrome finish for enhanced durability and readability
  • Comes complete with a protective vinyl case for convenient storage and transport