Learn More
MSC SPI Rectangular Steel Gage Block, Grade AS-1, 0.1004 in., 1 ea.
Catalog No. 5000516208
Encompass
This individual rectangular gage block is engineered from high-quality steel and manufactured to grade AS-1 specifications. Designed for precision metrology applications, this gage block provides reliable measurement standards for machine shops and quality control environments.
  • Features a 0.1004 in. size
  • Constructed from durable steel
  • Includes burr-proofed measuring surfaces
  • Designed with blended radii to prevent edge damage during use
  • Supplied with a certificate of calibration traceable to NIST
  • Suitable for stacking and wringing applications without damaging surfaces
Catalog No. 50-005-16208 Supplier MSC Supplier No. 87876702
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This individual rectangular gage block is engineered from high-quality steel and manufactured to grade AS-1 specifications. Designed for precision metrology applications, this gage block provides reliable measurement standards for machine shops and quality control environments.
  • Features a 0.1004 in. size
  • Constructed from durable steel
  • Includes burr-proofed measuring surfaces
  • Designed with blended radii to prevent edge damage during use
  • Supplied with a certificate of calibration traceable to NIST
  • Suitable for stacking and wringing applications without damaging surfaces