Learn More
MSC SPI Swiss Precision Instruments Rectangular Gage Block, Steel, Grade AS-1, 0.15000 in., 1 ea.
Catalog No. 5000511694
Encompass
This rectangular steel gage block is engineered for precision measurement applications, offering a size of 0.15000 in. and adhering to Grade AS-1 accuracy standards. The block is designed for durability and ease of use in metrology environments.
  • Constructed from steel for long-term stability
  • Features blended radii and a burr-proof design
  • Safe for wringing and stacking without damaging measuring surfaces
  • Includes a certificate of calibration traceable to NIST
  • Tolerance of + 0.000008 in./- 0.000004 in.
Catalog No. 50-005-11694 Supplier MSC Supplier No. 87877304
This rectangular steel gage block is engineered for precision measurement applications, offering a size of 0.15000 in. and adhering to Grade AS-1 accuracy standards. The block is designed for durability and ease of use in metrology environments.
  • Constructed from steel for long-term stability
  • Features blended radii and a burr-proof design
  • Safe for wringing and stacking without damaging measuring surfaces
  • Includes a certificate of calibration traceable to NIST
  • Tolerance of + 0.000008 in./- 0.000004 in.