Learn More
MSC SPI Rectangle Steel Gage Block, 0.10030 inch, Grade AS-1
Catalog No. 5000537318
Encompass
This individual rectangular steel gage block is a precision measurement tool. It features a size of 0.10030 inch and an AS-1 grade classification. Each block is designed with blended radii and is burr-proof, ensuring durability and measurement integrity during wringing or stacking.
  • Designed with blended radii and burr-proofed to prevent damage to measuring surfaces during wringing or stacking.
  • Includes a certificate of calibration traceable to NIST, ensuring accuracy and quality.
  • Made from steel for durability.
  • Features an AS-1 accuracy grade.
  • Has a tolerance of +0.000008/-0.000004 "
Catalog No. 50-005-37318 Supplier MSC Supplier No. 86291036
This individual rectangular steel gage block is a precision measurement tool. It features a size of 0.10030 inch and an AS-1 grade classification. Each block is designed with blended radii and is burr-proof, ensuring durability and measurement integrity during wringing or stacking.
  • Designed with blended radii and burr-proofed to prevent damage to measuring surfaces during wringing or stacking.
  • Includes a certificate of calibration traceable to NIST, ensuring accuracy and quality.
  • Made from steel for durability.
  • Features an AS-1 accuracy grade.
  • Has a tolerance of +0.000008/-0.000004 "