Learn More
MSC SPI Rectangle Steel Gage Block, 0.112 in, Grade 0, NIST Certified
Catalog No. 5000516524
Encompass
This gage block is a rectangular steel precision measurement tool designed for accuracy. It features a size of 0.112 inches and an accuracy grade of Grade 0, making it suitable for demanding calibration and inspection tasks. The block includes NIST Certification to ensure traceability and reliability.
  • Rectangular shape for versatile use
  • Grade 0 accuracy for high precision measurements
  • Made from durable steel for long-lasting performance
  • Precisely sized at 0.112 inches
  • Includes NIST certification for traceable accuracy
Catalog No. 50-005-16524 Supplier MSC Supplier No. 03266558
This gage block is a rectangular steel precision measurement tool designed for accuracy. It features a size of 0.112 inches and an accuracy grade of Grade 0, making it suitable for demanding calibration and inspection tasks. The block includes NIST Certification to ensure traceability and reliability.
  • Rectangular shape for versatile use
  • Grade 0 accuracy for high precision measurements
  • Made from durable steel for long-lasting performance
  • Precisely sized at 0.112 inches
  • Includes NIST certification for traceable accuracy