Learn More
MSC SPI Rectangle Steel Gage Block, 0.149 in, Grade 0, NIST Certified
Catalog No. 5000533366
Encompass
This gage block is a precision measuring tool, rectangular in shape and constructed from steel. It features an accuracy grade of 0 and a size of 0.149 " This product also includes NIST certification, ensuring its reliability for precise measurements.
  • Includes NIST certification
  • Rectangle shape
  • Grade 0 accuracy
  • 0.149 inch size
  • Steel material
Catalog No. 50-005-33366 Supplier MSC Supplier No. 03266921
This gage block is a precision measuring tool, rectangular in shape and constructed from steel. It features an accuracy grade of 0 and a size of 0.149 " This product also includes NIST certification, ensuring its reliability for precise measurements.
  • Includes NIST certification
  • Rectangle shape
  • Grade 0 accuracy
  • 0.149 inch size
  • Steel material