Learn More
MSC SPI Rectangle Steel Gage Block, 0.6 in, Grade 0, NIST Certified
Catalog No. 5000529776
Encompass
This SPI Rectangle Steel Gage Block is designed for precise measurement applications. It features a durable steel construction and offers a high level of accuracy, making it suitable for calibration and inspection tasks.
  • Constructed from steel
  • Rectangle shape
  • 0.6 in size
  • Grade 0 accuracy
  • Includes NIST certification
Catalog No. 50-005-29776 Supplier MSC Supplier No. 03267028
This SPI Rectangle Steel Gage Block is designed for precise measurement applications. It features a durable steel construction and offers a high level of accuracy, making it suitable for calibration and inspection tasks.
  • Constructed from steel
  • Rectangle shape
  • 0.6 in size
  • Grade 0 accuracy
  • Includes NIST certification