Learn More
MSC SPI Rectangular Steel Gage Block, 0.106 in, Grade AS-1, NIST Traceability Certified
Catalog No. 5000534652
Encompass
This rectangular steel gage block is designed for precision measurement applications. It features a specific size of 0.106 inch and meets Grade AS-1 accuracy standards, ensuring reliable performance. Each block includes NIST traceability certification, providing assurance of its metrological quality.
  • Precision rectangular steel gage block
  • 0.106 inch size for accurate measurements
  • Grade AS-1 accuracy for reliable performance
  • Includes NIST traceability certification
Catalog No. 50-005-34652 Supplier MSC Supplier No. 87876827
This rectangular steel gage block is designed for precision measurement applications. It features a specific size of 0.106 inch and meets Grade AS-1 accuracy standards, ensuring reliable performance. Each block includes NIST traceability certification, providing assurance of its metrological quality.
  • Precision rectangular steel gage block
  • 0.106 inch size for accurate measurements
  • Grade AS-1 accuracy for reliable performance
  • Includes NIST traceability certification