Learn More
MSC SPI Rectangular Steel Gage Block, 0.128 in, Grade AS-1, Steel
Catalog No. 5000530234
Encompass
This rectangular steel gage block is designed for high-precision measurement applications. It features blended radii and is burr-proof, ensuring accuracy and durability. The design prevents damage to measuring surfaces during wringing or stacking, and each block includes a Certificate of calibration traceable to NIST for assured quality.
  • Blended radii and burr-proof design
  • Wringing or stacking will not cause damage to measuring surfaces
  • Certificate of calibration traceable to NIST included
Catalog No. 50-005-30234 Supplier MSC Supplier No. 87877049
This rectangular steel gage block is designed for high-precision measurement applications. It features blended radii and is burr-proof, ensuring accuracy and durability. The design prevents damage to measuring surfaces during wringing or stacking, and each block includes a Certificate of calibration traceable to NIST for assured quality.
  • Blended radii and burr-proof design
  • Wringing or stacking will not cause damage to measuring surfaces
  • Certificate of calibration traceable to NIST included