Learn More
MSC SPI Scale Measurement Range Expander, Stainless Steel, Hole Depth 31.5 mm
Catalog No. 5000530869
Encompass
The SPI Scale Measurement Range Expander is an accessory designed to extend the measurement capabilities of a scale. Made from durable stainless steel, it features a 31.5 mm hole depth and is compatible with dial indicators to expand their measurement range.
  • Drop indicator accessory
  • Expands the measurement range of the scale
  • Use with dial indicator
  • Made from stainless steel
  • Features a 31.5 mm hole depth
Catalog No. 50-005-30869 Supplier MSC Supplier No. 77764769
The SPI Scale Measurement Range Expander is an accessory designed to extend the measurement capabilities of a scale. Made from durable stainless steel, it features a 31.5 mm hole depth and is compatible with dial indicators to expand their measurement range.
  • Drop indicator accessory
  • Expands the measurement range of the scale
  • Use with dial indicator
  • Made from stainless steel
  • Features a 31.5 mm hole depth