Learn More
MSC SPI Screw Pitch Gage, 16 Leaf, 1 to 12 TPI Range, Stainless Steel, 29° Thread Angle
Catalog No. 5000529360
Encompass
This screw pitch gage is designed for precise measurement of Acme inch threads. Featuring 16 leaves, it covers a threads per inch range of 1 to 12 and is constructed from durable stainless steel. The gage is suitable for both internal and external thread locations, and includes a locking mechanism for secure measurement.
  • Measurement type: Acme inch
  • Threads per inch range: 1 to 12
  • Thread angle: 29 degrees
  • Number of leaves: 16
  • Suitable for internal and external thread locations
  • Material: Stainless steel
  • Locking mechanism included
Catalog No. 50-005-29360 Supplier MSC Supplier No. 06377022
This screw pitch gage is designed for precise measurement of Acme inch threads. Featuring 16 leaves, it covers a threads per inch range of 1 to 12 and is constructed from durable stainless steel. The gage is suitable for both internal and external thread locations, and includes a locking mechanism for secure measurement.
  • Measurement type: Acme inch
  • Threads per inch range: 1 to 12
  • Thread angle: 29 degrees
  • Number of leaves: 16
  • Suitable for internal and external thread locations
  • Material: Stainless steel
  • Locking mechanism included