Learn More
MSC SPI Screw Pitch Gage, 20 Leaf, 0.4-6 mm Pitch Range, Steel
Catalog No. 5000513568
Encompass
The screw pitch gage is designed for precise metric measurements, featuring 20 leaves and a pitch range of 0.4 to 6.00 mm. Constructed from steel, it is suitable for both internal and external thread locations and includes a locking mechanism for secure use. This highly-rated gage is praised for its quality and ability to cover most common thread sizes.
  • Metric measurement type
  • Pitch range: 0.40-6.00 mm
  • Thread angle: 60.00 degrees
  • Number of leaves: 20
  • For internal and external thread locations
  • Steel material
  • Locking mechanism
  • Covers most common sizes
Catalog No. 50-005-13568 Supplier MSC Supplier No. 01221001
The screw pitch gage is designed for precise metric measurements, featuring 20 leaves and a pitch range of 0.4 to 6.00 mm. Constructed from steel, it is suitable for both internal and external thread locations and includes a locking mechanism for secure use. This highly-rated gage is praised for its quality and ability to cover most common thread sizes.
  • Metric measurement type
  • Pitch range: 0.40-6.00 mm
  • Thread angle: 60.00 degrees
  • Number of leaves: 20
  • For internal and external thread locations
  • Steel material
  • Locking mechanism
  • Covers most common sizes