Learn More
MSC SPI Screw Thread Micrometer, 3 to 4 in Range, Mechanical, 0.001 in Graduation
Catalog No. 5000528357
Encompass
This screw thread micrometer offers a 3 to 4 inch measuring range with a mechanical operation. It features a ratchet stop thimble for consistent pressure and a 0.001 inch graduation for precise readings. The micrometer also provides an accuracy of +/-0.0002 inch and includes interchangeable anvils, making it suitable for various thread measurement applications.
  • Mechanical operation for reliable measurements
  • 3 to 4 inch measuring range
  • Ratchet stop thimble ensures consistent measuring force
  • 0.001 inch graduation for high precision
  • Accuracy of +/-0.0002 inch
  • Interchangeable anvil type for versatility
  • Calibrated for assured accuracy
  • Durable painted finish
  • NIST traceability certification included
  • Comes with short form certificate of calibration, standards, and a protective case
Catalog No. 50-005-28357 Supplier MSC Supplier No. 37879517
This screw thread micrometer offers a 3 to 4 inch measuring range with a mechanical operation. It features a ratchet stop thimble for consistent pressure and a 0.001 inch graduation for precise readings. The micrometer also provides an accuracy of +/-0.0002 inch and includes interchangeable anvils, making it suitable for various thread measurement applications.
  • Mechanical operation for reliable measurements
  • 3 to 4 inch measuring range
  • Ratchet stop thimble ensures consistent measuring force
  • 0.001 inch graduation for high precision
  • Accuracy of +/-0.0002 inch
  • Interchangeable anvil type for versatility
  • Calibrated for assured accuracy
  • Durable painted finish
  • NIST traceability certification included
  • Comes with short form certificate of calibration, standards, and a protective case