Learn More
MSC SPI Setup Block, 0.0003 Squareness, Hardened Steel, 2-3-4 Block
Catalog No. 5000529638
Encompass
This setup block is a 2-3-4 configuration, constructed from hardened steel with a precise squareness of 0.0003 per 1 " It boasts a hardness rating of 55 to 60 RC, an overall length of 3 inches, and a 3/8-16 tapped hole size. Measuring 2 inches in overall width, it is sold as a matched pair and includes a 9-inch fluorescent clock.
  • 0.0003 squareness per 1 inch
  • Hardened steel construction
  • 55 to 60 RC hardness
  • 2-3-4 block type
  • Overall length of 3 inches
  • 3/8-16 tapped hole size
  • Overall width of 2 inches
  • Sold as a matched pair
  • Includes a 9 inch fluorescent clock
Catalog No. 50-005-29638 Supplier MSC Supplier No. 03433612
This setup block is a 2-3-4 configuration, constructed from hardened steel with a precise squareness of 0.0003 per 1 " It boasts a hardness rating of 55 to 60 RC, an overall length of 3 inches, and a 3/8-16 tapped hole size. Measuring 2 inches in overall width, it is sold as a matched pair and includes a 9-inch fluorescent clock.
  • 0.0003 squareness per 1 inch
  • Hardened steel construction
  • 55 to 60 RC hardness
  • 2-3-4 block type
  • Overall length of 3 inches
  • 3/8-16 tapped hole size
  • Overall width of 2 inches
  • Sold as a matched pair
  • Includes a 9 inch fluorescent clock