Learn More
MSC SPI Setup Block Set, 1-2-3 & 2-4-6 Block Set, Hardened Steel
Catalog No. 5000531312
Encompass
This product is a versatile set of setup blocks designed for various applications, comprising both 1-2-3 and 2-4-6 blocks. Constructed from hardened steel, these blocks are built for durability and precision in industrial settings.
  • Includes one pair of 1-2-3 blocks and one pair of 2-4-6 blocks
  • Manufactured from hardened steel for enhanced durability
  • Features tapped holes for versatile mounting and securing options
  • Offers squareness of 0.0003 and 0.0001 per 1 inch
  • Each pair of blocks comes with its own protective case
Catalog No. 50-005-31312 Supplier MSC Supplier No. 86252855
This product is a versatile set of setup blocks designed for various applications, comprising both 1-2-3 and 2-4-6 blocks. Constructed from hardened steel, these blocks are built for durability and precision in industrial settings.
  • Includes one pair of 1-2-3 blocks and one pair of 2-4-6 blocks
  • Manufactured from hardened steel for enhanced durability
  • Features tapped holes for versatile mounting and securing options
  • Offers squareness of 0.0003 and 0.0001 per 1 inch
  • Each pair of blocks comes with its own protective case