Learn More
MSC SPI Small Hole Gage, 0.3 to 0.4 in Measurement, 1-9/16 in Gage Depth
Catalog No. 5000511373
Encompass
This small hole gage is designed for precise measurement within a range of 0.3 to 0.4 in. It features a 1-9/16 in gage depth and a 3 in overall length, complete with a satin chrome finish and full ball style, making it suitable for various measurement applications.
  • Measures holes within a 0.3 to 0.4 in range.
  • Features a 1-9/16 in gage depth.
  • Overall length of 3 in for comfortable handling.
  • Durable satin chrome finish.
  • Equipped with a full ball style for accurate readings.
  • Manufactured in Japan.
Catalog No. 50-005-11373 Supplier MSC Supplier No. 01719236
This small hole gage is designed for precise measurement within a range of 0.3 to 0.4 in. It features a 1-9/16 in gage depth and a 3 in overall length, complete with a satin chrome finish and full ball style, making it suitable for various measurement applications.
  • Measures holes within a 0.3 to 0.4 in range.
  • Features a 1-9/16 in gage depth.
  • Overall length of 3 in for comfortable handling.
  • Durable satin chrome finish.
  • Equipped with a full ball style for accurate readings.
  • Manufactured in Japan.