Learn More
MSC SPI Spare Part Kit, For 21-337-1 & 21-344-7 Dial Bore Gauge, Includes #2 Anvil & #3 Washer Sets
Catalog No. 5000532696
Encompass
This spare part kit is designed for bore gage accessories, specifically for use with 21-337-1 and 21-344-7 Dial Bore Gauges. It provides essential components for maintenance and repair of these specific gauges.
  • Spare part kit for bore gage accessories
  • Designed for use with 21-337-1 and 21-344-7 dial bore gauges
  • Includes #2 anvil and #3 washer sets
  • Contains 2 pieces
Catalog No. 50-005-32696 Supplier MSC Supplier No. 39441175
This spare part kit is designed for bore gage accessories, specifically for use with 21-337-1 and 21-344-7 Dial Bore Gauges. It provides essential components for maintenance and repair of these specific gauges.
  • Spare part kit for bore gage accessories
  • Designed for use with 21-337-1 and 21-344-7 dial bore gauges
  • Includes #2 anvil and #3 washer sets
  • Contains 2 pieces