Learn More
MSC SPI Spare Part Kit, For 21-376-9 Series Test Indicator, 5 Piece
Catalog No. 5000514071
Encompass
This spare part kit is designed for the 21-376-9 series test indicator, providing essential components for maintenance and repair. It offers a convenient solution for replacing commonly used parts, ensuring the continued accuracy and functionality of the indicator.
  • Accessory type: Spare part kit
  • For use with 21-376-9 series test indicator
  • Country of origin: China
  • Quantity: 5 piece kit
  • Kit contents include: bezel, 1 1/4 crystal, 5/32 in stem, 3/8 in stem, and wrench
Catalog No. 50-005-14071 Supplier MSC Supplier No. 39441241
This spare part kit is designed for the 21-376-9 series test indicator, providing essential components for maintenance and repair. It offers a convenient solution for replacing commonly used parts, ensuring the continued accuracy and functionality of the indicator.
  • Accessory type: Spare part kit
  • For use with 21-376-9 series test indicator
  • Country of origin: China
  • Quantity: 5 piece kit
  • Kit contents include: bezel, 1 1/4 crystal, 5/32 in stem, 3/8 in stem, and wrench