Learn More
MSC SPI Square Set, 4 Piece, Hardened Steel, Beveled Squares
Catalog No. 5000529446
Encompass
This 4-piece square set is crafted from hardened steel, designed for precision measurement and layout tasks. It includes three beveled squares, a protective case, and a triangular straight edge, providing essential tools for demanding applications.
  • Crafted from hardened steel for durability and precision
  • Includes three beveled squares for accurate angle checks
  • Comes with a protective case for convenient storage and transport
  • Features a triangular straight edge for precise line drawing
  • Hardened and ground construction ensures enhanced accuracy and longevity
  • Offers a range of base lengths (3/4, 1-1/8, 1-15/16 in) and blade lengths (1, 1-1/2, 1-9/16, 2-15/16 in) for versatile use
Catalog No. 50-005-29446 Supplier MSC Supplier No. 06506505
This 4-piece square set is crafted from hardened steel, designed for precision measurement and layout tasks. It includes three beveled squares, a protective case, and a triangular straight edge, providing essential tools for demanding applications.
  • Crafted from hardened steel for durability and precision
  • Includes three beveled squares for accurate angle checks
  • Comes with a protective case for convenient storage and transport
  • Features a triangular straight edge for precise line drawing
  • Hardened and ground construction ensures enhanced accuracy and longevity
  • Offers a range of base lengths (3/4, 1-1/8, 1-15/16 in) and blade lengths (1, 1-1/2, 1-9/16, 2-15/16 in) for versatile use