Learn More
MSC SPI Steel Gage Block, 0.1007 in, Grade 0
Catalog No. 5000511442
Encompass
This rectangular steel gage block provides precise measurements for calibration and inspection tasks. It features a size of 0.1007 inches and an accuracy grade of 0, ensuring high precision for demanding applications.
  • Includes NIST traceable calibration certificate.
  • Each gage block has blended radii and is burr-proofed.
  • Wringing or stacking the block will not cause damage to the measuring surfaces.
Catalog No. 50-005-11442 Supplier MSC Supplier No. 03266418
This rectangular steel gage block provides precise measurements for calibration and inspection tasks. It features a size of 0.1007 inches and an accuracy grade of 0, ensuring high precision for demanding applications.
  • Includes NIST traceable calibration certificate.
  • Each gage block has blended radii and is burr-proofed.
  • Wringing or stacking the block will not cause damage to the measuring surfaces.