Learn More
MSC SPI Steel Rule, 12 in OAL, 16R Graduation, 1 in OAW
Catalog No. 5000530352
Encompass
This steel rule is crafted from durable tool steel and features a satin chrome-plated finish. It is 12 inches in overall length with a 1 inch overall width, offering precise measurements with a 16R graduation style and multiple fine graduations for versatile use.
  • Crafted from tool steel for durability
  • 12 inch overall length
  • 16R graduation style
  • Features 1/64, 1/100, 1/32, and 1/50 inch graduations
  • Silver color
  • Includes hook for convenient use
  • Not magnetic
  • Horizontal scale orientation
  • 1 inch overall width
  • 0.0400 inch thickness
  • Satin chrome coating/finish
Catalog No. 50-005-30352 Supplier MSC Supplier No. 69745784
This steel rule is crafted from durable tool steel and features a satin chrome-plated finish. It is 12 inches in overall length with a 1 inch overall width, offering precise measurements with a 16R graduation style and multiple fine graduations for versatile use.
  • Crafted from tool steel for durability
  • 12 inch overall length
  • 16R graduation style
  • Features 1/64, 1/100, 1/32, and 1/50 inch graduations
  • Silver color
  • Includes hook for convenient use
  • Not magnetic
  • Horizontal scale orientation
  • 1 inch overall width
  • 0.0400 inch thickness
  • Satin chrome coating/finish