Learn More
MSC SPI Steel Rule, 18 in, 4R Graduation, Tool Steel, Satin Chrome-Plated
Catalog No. 5000534471
Encompass
The SPI steel rule provides precise measurement capabilities for various applications. Constructed from durable tool steel with a satin chrome-plated finish, it is designed for accuracy and longevity. This rule features a convenient hook and clear, multi-scale graduations to ensure reliable readings.
  • Overall length of 18 inch
  • 4R graduation, including 1/16, 1/32, 1/64, and 1/8 inch increments
  • Overall width of 1-1/8 inch
  • Constructed from tool steel
  • Silver, satin chrome-plated finish
  • Includes a hook for easy use
  • Horizontal scale orientation
  • Thickness of 0.0400 inch
Catalog No. 50-005-34471 Supplier MSC Supplier No. 69745800
The SPI steel rule provides precise measurement capabilities for various applications. Constructed from durable tool steel with a satin chrome-plated finish, it is designed for accuracy and longevity. This rule features a convenient hook and clear, multi-scale graduations to ensure reliable readings.
  • Overall length of 18 inch
  • 4R graduation, including 1/16, 1/32, 1/64, and 1/8 inch increments
  • Overall width of 1-1/8 inch
  • Constructed from tool steel
  • Silver, satin chrome-plated finish
  • Includes a hook for easy use
  • Horizontal scale orientation
  • Thickness of 0.0400 inch