Learn More
MSC SPI Steel Rule, 18 in, 5R Graduation, Rigid, Satin Chrome
Catalog No. 5000531781
Encompass
This steel rule is designed for precision measurements, featuring an 18 inch overall length, 5R graduation, and a rigid construction. Its satin chrome finish enhances readability and durability for various applications.
  • 18 inch overall length.
  • 5R graduation style.
  • Rigid flexibility.
  • Satin chrome coating/finish.
  • Constructed from steel.
  • Graduations in 1/64, 1/100, and 1/32 "
  • 1-1/8 inch overall width.
  • 0.0400 inch thickness.
Catalog No. 50-005-31781 Supplier MSC Supplier No. 59624759
This steel rule is designed for precision measurements, featuring an 18 inch overall length, 5R graduation, and a rigid construction. Its satin chrome finish enhances readability and durability for various applications.
  • 18 inch overall length.
  • 5R graduation style.
  • Rigid flexibility.
  • Satin chrome coating/finish.
  • Constructed from steel.
  • Graduations in 1/64, 1/100, and 1/32 "
  • 1-1/8 inch overall width.
  • 0.0400 inch thickness.