Learn More
MSC SPI Steel Rule, 6 inch, Decimal/Metric Graduation, Rigid, Satin Chrome
Catalog No. 5000531632
Encompass
This steel rule has a 6 inch overall length and features both decimal and metric graduations, making it versatile for various measurements. Its rigid design ensures stability during use, and the satin chrome finish provides durability and clear readability. It is suitable for applications requiring precise linear measurements.
  • Constructed from durable steel
  • Features a 6 inch (150 mm) length for general measurement tasks
  • Offers both decimal and metric graduation styles
  • Designed with a rigid body for stable use
  • Measures 3/4 inch in width
  • Finished with satin chrome for enhanced durability and visibility
Catalog No. 50-005-31632 Supplier MSC Supplier No. 76417955
This steel rule has a 6 inch overall length and features both decimal and metric graduations, making it versatile for various measurements. Its rigid design ensures stability during use, and the satin chrome finish provides durability and clear readability. It is suitable for applications requiring precise linear measurements.
  • Constructed from durable steel
  • Features a 6 inch (150 mm) length for general measurement tasks
  • Offers both decimal and metric graduation styles
  • Designed with a rigid body for stable use
  • Measures 3/4 inch in width
  • Finished with satin chrome for enhanced durability and visibility