Learn More
MSC SPI Steel Rule, 6 in OAL, 16R Graduation, Flexible, Satin Chrome-Plated
Catalog No. 5000529311
Encompass
This flexible steel rule features a 6 inch overall length and 16R graduation. It has a satin chrome-plated finish and includes graduations of 1/100, 1/32, 1/50, and 1/64 inches. Constructed from steel, it has a thickness of 0.0150 "
  • Flexible design
  • 6 inch overall length
  • 16R graduation style
  • Satin chrome finish
  • Includes 1/100, 1/32, 1/50, and 1/64 inch graduations
  • Constructed from steel
  • 0.0150 inch thickness
  • Does not include a hook
  • Not magnetic
Catalog No. 50-005-29311 Supplier MSC Supplier No. 42614776
This flexible steel rule features a 6 inch overall length and 16R graduation. It has a satin chrome-plated finish and includes graduations of 1/100, 1/32, 1/50, and 1/64 inches. Constructed from steel, it has a thickness of 0.0150 "
  • Flexible design
  • 6 inch overall length
  • 16R graduation style
  • Satin chrome finish
  • Includes 1/100, 1/32, 1/50, and 1/64 inch graduations
  • Constructed from steel
  • 0.0150 inch thickness
  • Does not include a hook
  • Not magnetic