Learn More
MSC SPI Surface Roughness Gage, Rt, Rz, Ra, Rq, 5 μm Stylus Tip
Catalog No. 5000513397
Encompass
The SPI Surface Roughness Gage is designed for precise measurement of surface finish. It is capable of evaluating multiple roughness parameters, making it suitable for a wide range of industrial and laboratory applications.
  • Measures Rt, Rz, Ra, and Rq roughness parameters.
  • Equipped with a 5 μm stylus tip radius.
  • Features an LCD display for clear readings.
  • Includes Bluetooth for data output.
  • Powered by an A/C rechargeable lithium-ion battery.
  • Comes with one battery included.
  • Operates within a temperature range of 0-40°C (32-105°F).
Catalog No. 50-005-13397 Supplier MSC Supplier No. 05228671
The SPI Surface Roughness Gage is designed for precise measurement of surface finish. It is capable of evaluating multiple roughness parameters, making it suitable for a wide range of industrial and laboratory applications.
  • Measures Rt, Rz, Ra, and Rq roughness parameters.
  • Equipped with a 5 μm stylus tip radius.
  • Features an LCD display for clear readings.
  • Includes Bluetooth for data output.
  • Powered by an A/C rechargeable lithium-ion battery.
  • Comes with one battery included.
  • Operates within a temperature range of 0-40°C (32-105°F).