Learn More
MSC SPI Swiss Precision Instruments Gage Block, 0.1008 in, Steel, Grade 0
Catalog No. 5000529651
Encompass
This precision rectangular gage block from SPI Swiss Precision Instruments is designed for accurate measurement and calibration. Made from durable steel, it features a 0.1008 inch size and an accuracy grade of 0, ensuring high precision for demanding applications. It also includes NIST traceability certification.
  • Rectangular form
  • Made from steel
  • 0.1008 inch size
  • Accuracy grade 0
  • Includes NIST traceability certification
Catalog No. 50-005-29651 Supplier MSC Supplier No. 03266426
This precision rectangular gage block from SPI Swiss Precision Instruments is designed for accurate measurement and calibration. Made from durable steel, it features a 0.1008 inch size and an accuracy grade of 0, ensuring high precision for demanding applications. It also includes NIST traceability certification.
  • Rectangular form
  • Made from steel
  • 0.1008 inch size
  • Accuracy grade 0
  • Includes NIST traceability certification