Learn More
MSC SPI Swiss Precision Instruments Plug Gage, Class ZZ Plus, 0.5450 in, 0.0002 in
Catalog No. 5000516340
Encompass
The SPI Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate diameter checks. It features a 0.5450 in diameter and a tight 0.0002 in tolerance, making it suitable for demanding applications. Manufactured from durable steel, it ensures reliability and longevity in various industrial environments.
  • Designed for precise measurement of bore diameters.
  • Features a 0.5450 in measuring diameter.
  • Provides a tight 0.0002 in tolerance for accuracy.
  • Constructed from durable steel for extended life.
  • Classified as Class ZZ Plus for specific application requirements.
Catalog No. 50-005-16340 Supplier MSC Supplier No. 78387305
The SPI Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate diameter checks. It features a 0.5450 in diameter and a tight 0.0002 in tolerance, making it suitable for demanding applications. Manufactured from durable steel, it ensures reliability and longevity in various industrial environments.
  • Designed for precise measurement of bore diameters.
  • Features a 0.5450 in measuring diameter.
  • Provides a tight 0.0002 in tolerance for accuracy.
  • Constructed from durable steel for extended life.
  • Classified as Class ZZ Plus for specific application requirements.