Learn More
MSC SPI Swiss Precision Instruments Plug & Pin Gage, 0.246 in Diameter, Steel, 0.0002 in Tolerance
Catalog No. 5000528704
Encompass
The SPI Swiss Precision Instruments Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate diameter checks. Made from steel, it features a 0.246 inch diameter and a tight tolerance of 0.0002 inch, ensuring high precision for various applications.
  • Designed for precise diameter checks.
  • Made from durable steel material.
  • Features a 0.246 inch diameter.
  • Maintains a tight 0.0002 inch tolerance.
  • Classified as Class ZZ Minus for specific gauging requirements.
Catalog No. 50-005-28704 Supplier MSC Supplier No. 56245954
Add to Cart
Add to Cart
The SPI Swiss Precision Instruments Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate diameter checks. Made from steel, it features a 0.246 inch diameter and a tight tolerance of 0.0002 inch, ensuring high precision for various applications.
  • Designed for precise diameter checks.
  • Made from durable steel material.
  • Features a 0.246 inch diameter.
  • Maintains a tight 0.0002 inch tolerance.
  • Classified as Class ZZ Minus for specific gauging requirements.