Learn More
MSC SPI Swiss Precision Instruments Plug & Pin Gage, 0.477 in, ZZ Minus, Black Oxide
Catalog No. 5000531808
Encompass
This plug and pin gage is designed for precision measurement applications. It features a specific diameter, a Class ZZ Minus tolerance, and a black oxide finish for durability.
  • 0.477 in diameter
  • Class ZZ Minus tolerance
  • Black oxide finish
  • Suitable for precision measurement applications
Catalog No. 50-005-31808 Supplier MSC Supplier No. 56248263
This plug and pin gage is designed for precision measurement applications. It features a specific diameter, a Class ZZ Minus tolerance, and a black oxide finish for durability.
  • 0.477 in diameter
  • Class ZZ Minus tolerance
  • Black oxide finish
  • Suitable for precision measurement applications