Learn More
MSC SPI Swiss Precision Instruments Plug & Pin Gage, Class ZZ Minus, 0.730 in Dia, Black Oxide
Catalog No. 5000530112
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.730 inch diameter and a durable black oxide finish, making it suitable for a variety of industrial and laboratory uses.
  • Designed for precise measurement applications
  • Diameter of 0.730 inches
  • Tolerance of 0.000200 inches
  • Black oxide finish for durability
Catalog No. 50-005-30112 Supplier MSC Supplier No. 56250798
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.730 inch diameter and a durable black oxide finish, making it suitable for a variety of industrial and laboratory uses.
  • Designed for precise measurement applications
  • Diameter of 0.730 inches
  • Tolerance of 0.000200 inches
  • Black oxide finish for durability