Learn More
MSC SPI Taperlock Thread Plug Gage, 5/16-18, 2B Class, Double End, Go/No Go
Catalog No. 5000536057
Encompass
This Taperlock thread plug gage is designed to measure the correctness of threads. It features a double-end design with both "Go" and "No Go" ends, ensuring precise thread verification. The gage comes with a certificate traceable to NIST, and thread measurements adhere to ANSI specification 1.16B.
  • Measures threads for correctness
  • "Go" end verifies proper threading
  • "No Go" end checks thread limits
  • Constructed from steel
  • Includes a handle for ease of use
  • NIST traceable certification for accuracy
  • Measures according to ANSI specification 1.16B
Catalog No. 50-005-36057 Supplier MSC Supplier No. 75889311
This Taperlock thread plug gage is designed to measure the correctness of threads. It features a double-end design with both "Go" and "No Go" ends, ensuring precise thread verification. The gage comes with a certificate traceable to NIST, and thread measurements adhere to ANSI specification 1.16B.
  • Measures threads for correctness
  • "Go" end verifies proper threading
  • "No Go" end checks thread limits
  • Constructed from steel
  • Includes a handle for ease of use
  • NIST traceable certification for accuracy
  • Measures according to ANSI specification 1.16B