Learn More
MSC SPI Telescoping Gage, 3-1/2-6 in, 12 in Length, Hardened Tool Steel, Satin Chrome Finish
Catalog No. 5000534755
Encompass
The SPI Telescoping Gage is designed for precise measurements. It features a range of 3-1/2-6 in and an overall length of 12 in. Constructed from hardened tool steel with a satin chrome finish, this gage is built for durability and accuracy.
  • 12 in long handle
  • Hardened tool steel construction
  • Satin chrome finish
  • 3-1/2-6 in measuring range
  • 12 in overall length
Catalog No. 50-005-34755 Supplier MSC Supplier No. 73896706
The SPI Telescoping Gage is designed for precise measurements. It features a range of 3-1/2-6 in and an overall length of 12 in. Constructed from hardened tool steel with a satin chrome finish, this gage is built for durability and accuracy.
  • 12 in long handle
  • Hardened tool steel construction
  • Satin chrome finish
  • 3-1/2-6 in measuring range
  • 12 in overall length