Learn More
MSC SPI Telescoping Gage, 5/16 to 1/2 Inch Range, 5-7/8 Inch Overall Length, Hardened Tool Steel
Catalog No. 5000536238
Encompass
This telescoping gage is designed for precise internal measurements within a range of 5/16 to 1/2 " It features a durable construction and is suitable for various industrial applications.
  • Has a 4 inch long handle.
  • Constructed from hardened tool steel.
  • Features a satin chrome finish.
Catalog No. 50-005-36238 Supplier MSC Supplier No. 09232570
This telescoping gage is designed for precise internal measurements within a range of 5/16 to 1/2 " It features a durable construction and is suitable for various industrial applications.
  • Has a 4 inch long handle.
  • Constructed from hardened tool steel.
  • Features a satin chrome finish.