Learn More
MSC SPI Test Indicator Contact Point, 0.0390 in Ball Diameter, Steel
Catalog No. 5000533571
Encompass
This test indicator contact point features a 0.0390 in (1.00 mm) ball diameter and is constructed from steel. It is specifically designed for precise measurements when used with Peacock Test Indicators.
  • Ball diameter of 0.0390 in (1.00 mm)
  • Made from steel
  • Designed for use with test indicators
  • Ball style contact point
Catalog No. 50-005-33571 Supplier MSC Supplier No. 35893825
This test indicator contact point features a 0.0390 in (1.00 mm) ball diameter and is constructed from steel. It is specifically designed for precise measurements when used with Peacock Test Indicators.
  • Ball diameter of 0.0390 in (1.00 mm)
  • Made from steel
  • Designed for use with test indicators
  • Ball style contact point