Learn More
MSC SPI Test Indicator Contact Point, 0.0780 in Ball Diameter, Steel
Catalog No. 5000514105
Encompass
This test indicator contact point features a 0.0780 inch steel ball diameter, providing precise measurements for various applications.
  • Ball style.
  • Features a 0.0780 inch ball diameter.
  • Made from steel contact point material.
  • Designed for use with test indicators.
  • Manufactured in the United States.
Catalog No. 50-005-14105 Supplier MSC Supplier No. 35893817
This test indicator contact point features a 0.0780 inch steel ball diameter, providing precise measurements for various applications.
  • Ball style.
  • Features a 0.0780 inch ball diameter.
  • Made from steel contact point material.
  • Designed for use with test indicators.
  • Manufactured in the United States.