Learn More
MSC SPI Test Indicator Contact Point, 0.0800 in Ball Diameter, Carbide
Catalog No. 5000529505
Encompass
This test indicator contact point features a 0.0800 inch ball diameter and is made from carbide, designed for use with test indicators.
  • Ball style
  • 0.0800 inch ball diameter
  • 0.0800 inch tip diameter
  • Carbide contact point material
  • Suitable for use with test indicators
Catalog No. 50-005-29505 Supplier MSC Supplier No. 06445233
This test indicator contact point features a 0.0800 inch ball diameter and is made from carbide, designed for use with test indicators.
  • Ball style
  • 0.0800 inch ball diameter
  • 0.0800 inch tip diameter
  • Carbide contact point material
  • Suitable for use with test indicators