Learn More
MSC SPI Test Indicator Contact Point, 0.1000 in Ball Diameter, Carbide, 2-3/4 in Contact Point Length
Catalog No. 5000529539
Encompass
This Test Indicator Contact Point features a 0.1000 in ball diameter and a 2-3/4 in contact point length. Made from durable carbide, it is designed for use with test indicators, ensuring compatibility and reliable performance in measuring and inspecting applications.
  • 0.1000 in ball diameter
  • Durable carbide material
  • 2-3/4 in contact point length
  • For use with test indicators
Catalog No. 50-005-29539 Supplier MSC Supplier No. 06445357
This Test Indicator Contact Point features a 0.1000 in ball diameter and a 2-3/4 in contact point length. Made from durable carbide, it is designed for use with test indicators, ensuring compatibility and reliable performance in measuring and inspecting applications.
  • 0.1000 in ball diameter
  • Durable carbide material
  • 2-3/4 in contact point length
  • For use with test indicators