Learn More
MSC SPI Test Indicator Kit, 10 Pc, 0.060 in Max, 1.180 in Dial Dia.
Catalog No. 5000528343
Encompass
This Test Indicator Kit provides precise measurements for various metrology applications. It features a horizontal indicator style and includes 10 pieces, offering versatility and accuracy for professionals.
  • Automatic reversal for easy readings.
  • Dovetail stems for flexible mounting.
  • Jeweled movement ensures smooth operation and durability.
  • Revolution counter for accurate tracking of full rotations.
  • Rotating dial allows for precise zero setting.
  • Calibrated for assured accuracy.
  • Traceability certification included for quality assurance.
  • Includes stems, contact points, and other essential accessories for comprehensive use.
Catalog No. 50-005-28343 Supplier MSC Supplier No. 37543998
This Test Indicator Kit provides precise measurements for various metrology applications. It features a horizontal indicator style and includes 10 pieces, offering versatility and accuracy for professionals.
  • Automatic reversal for easy readings.
  • Dovetail stems for flexible mounting.
  • Jeweled movement ensures smooth operation and durability.
  • Revolution counter for accurate tracking of full rotations.
  • Rotating dial allows for precise zero setting.
  • Calibrated for assured accuracy.
  • Traceability certification included for quality assurance.
  • Includes stems, contact points, and other essential accessories for comprehensive use.