Learn More
MSC SPI Test Indicator Kit, 8 Pc, 0.008 in Max, 0.0001 in Accuracy
Catalog No. 5000528356
Encompass
This test indicator kit provides precise measurements with an 0.008 inch maximum range and 0.0001 inch accuracy. It features an easy-to-read dial and universal positioning capability, suitable for various measurement tasks.
  • Easy-to-read dial
  • Universal positioning capability
  • Jeweled bearing for smooth operation
  • Supplied with 3/8 inch dovetail stems
  • Short form calibration certificate included
  • Furnished in a fitted case for protection and organization
  • Includes indicator, two contact points, universal clamp, and height gage bar
  • 3/8 inch and 5/32 inch dovetail extensions for inch sets are included
Catalog No. 50-005-28356 Supplier MSC Supplier No. 37544079
This test indicator kit provides precise measurements with an 0.008 inch maximum range and 0.0001 inch accuracy. It features an easy-to-read dial and universal positioning capability, suitable for various measurement tasks.
  • Easy-to-read dial
  • Universal positioning capability
  • Jeweled bearing for smooth operation
  • Supplied with 3/8 inch dovetail stems
  • Short form calibration certificate included
  • Furnished in a fitted case for protection and organization
  • Includes indicator, two contact points, universal clamp, and height gage bar
  • 3/8 inch and 5/32 inch dovetail extensions for inch sets are included