Learn More
MSC SPI Test Indicator Kit, 9 Pc, 0.2500" Max, 0.0002" Accuracy
Catalog No. 5000513239
Encompass
This 9-piece Test Indicator Kit provides precise measurements for various applications. It features a maximum measurement of 0.2500 inches and an accuracy of 0.0002 inches, making it suitable for demanding laboratory and industrial environments. The kit includes both a stationary face indicator and a centering indicator, along with various feelers, all housed in a fitted case for protection and portability.
  • Includes stationary face indicator and centering indicator
  • Maximum measurement of 0.2500 in
  • Accuracy of 0.0002 in
  • Dial graduation of 0.000500 in
  • Fitted case for organized storage and transport
  • Contains three internal feelers, a restraining arm, a center point feele, and three external feelers
  • White dial for clear readings
Catalog No. 50-005-13239 Supplier MSC Supplier No. 09232760
This 9-piece Test Indicator Kit provides precise measurements for various applications. It features a maximum measurement of 0.2500 inches and an accuracy of 0.0002 inches, making it suitable for demanding laboratory and industrial environments. The kit includes both a stationary face indicator and a centering indicator, along with various feelers, all housed in a fitted case for protection and portability.
  • Includes stationary face indicator and centering indicator
  • Maximum measurement of 0.2500 in
  • Accuracy of 0.0002 in
  • Dial graduation of 0.000500 in
  • Fitted case for organized storage and transport
  • Contains three internal feelers, a restraining arm, a center point feele, and three external feelers
  • White dial for clear readings