Learn More
MSC SPI Testing File, Rockwell C Scale, 65 RC
Catalog No. 5000516994
Encompass
This testing file is designed for use with a hardness tester. It provides a reliable method for verifying the hardness of materials.
  • For use with hardness testers
  • Rockwell C scale type
  • Maximum hardness of 65 RC
Catalog No. 50-005-16994 Supplier MSC Supplier No. 00692525
This testing file is designed for use with a hardness tester. It provides a reliable method for verifying the hardness of materials.
  • For use with hardness testers
  • Rockwell C scale type
  • Maximum hardness of 65 RC