Learn More
MSC SPI Plug Thread Gage, 1/2-20, 2B Class, Go, 1 ea
Catalog No. 5000511887
Encompass
This single-end taperlock thread plug gage is designed for the inspection of internal threads to ensure the proper assembly of mating components. Manufactured from tool steel, this gage adheres to ASME B1.2 standards and follows ANSI specification 1.16B for thread measurements.
  • Thread size of 1/2-20 UNF
  • Class 2B classification
  • Single end GO configuration
  • Inspection certificate traceable to NIST included
  • Overall length of 1.7500 in. (44.45 mm)
  • Right hand thread direction
  • Taperlock design
Catalog No. 50-005-11887 Supplier MSC Supplier No. 75888388
This single-end taperlock thread plug gage is designed for the inspection of internal threads to ensure the proper assembly of mating components. Manufactured from tool steel, this gage adheres to ASME B1.2 standards and follows ANSI specification 1.16B for thread measurements.
  • Thread size of 1/2-20 UNF
  • Class 2B classification
  • Single end GO configuration
  • Inspection certificate traceable to NIST included
  • Overall length of 1.7500 in. (44.45 mm)
  • Right hand thread direction
  • Taperlock design