Learn More
MSC SPI Thread Ring Gage, M14 x 2, Metric, Class 6G, Go
Catalog No. 5000530992
Encompass
This "Go" style metric thread ring gage is designed for the precise inspection of M14 x 2 external threads. It features a 6G class fit and complies with ISO 1502 standards, ensuring accuracy and reliability in thread verification. Each gage is supplied with a Certificate of Calibration traceable to the National Physical Laboratory (NPL).
  • "Go" style gage for external thread inspection.
  • M14 x 2 thread size.
  • Metric thread type.
  • Class 6G fit for precise measurements.
  • Manufactured from durable tool steel.
  • Conforms to ISO 1502 thread dimensions.
  • Includes a certificate of calibration traceable to NPL standards.
Catalog No. 50-005-30992 Supplier MSC Supplier No. 75890467
This "Go" style metric thread ring gage is designed for the precise inspection of M14 x 2 external threads. It features a 6G class fit and complies with ISO 1502 standards, ensuring accuracy and reliability in thread verification. Each gage is supplied with a Certificate of Calibration traceable to the National Physical Laboratory (NPL).
  • "Go" style gage for external thread inspection.
  • M14 x 2 thread size.
  • Metric thread type.
  • Class 6G fit for precise measurements.
  • Manufactured from durable tool steel.
  • Conforms to ISO 1502 thread dimensions.
  • Includes a certificate of calibration traceable to NPL standards.