Learn More
MSC SPI Threaded Ring Gage, 3/4-16, Class 2A, Go & No Go
Catalog No. 5000528224
Encompass
This adjustable threaded ring gage is designed for the inspection of threads, ensuring proper assembly of mating components. It features both Go and No Go ends, a 3/4-16 UNF thread size, and a 2A classification, constructed from durable tool steel.
  • Adjustable type for precise inspection.
  • Ensures proper assembly of mating components.
  • Features both Go and No Go ends.
  • Designed for 3/4-16 UNF thread size.
  • Class 2A classification.
  • Constructed from durable tool steel.
  • Double ring design.
  • Meets ASME B1.2 and ANSI standards.
  • Includes inspection certificate traceable to NIST.
  • Right hand thread direction.
Catalog No. 50-005-28224 Supplier MSC Supplier No. 46553913
This adjustable threaded ring gage is designed for the inspection of threads, ensuring proper assembly of mating components. It features both Go and No Go ends, a 3/4-16 UNF thread size, and a 2A classification, constructed from durable tool steel.
  • Adjustable type for precise inspection.
  • Ensures proper assembly of mating components.
  • Features both Go and No Go ends.
  • Designed for 3/4-16 UNF thread size.
  • Class 2A classification.
  • Constructed from durable tool steel.
  • Double ring design.
  • Meets ASME B1.2 and ANSI standards.
  • Includes inspection certificate traceable to NIST.
  • Right hand thread direction.