Learn More
MSC SPI Threaded Ring Gage, 5/8-11 Thread, Class 2A, Go/No Go
Catalog No. 5000536758
Encompass
This threaded ring gage is designed for precise inspection of threads to ensure the proper assembly of mating components. It features an adjustable type for versatile use in various applications.
  • Adjustable design for flexible application
  • Ensures proper assembly of mating components through accurate thread inspection
  • Includes both Go and No Go functions for comprehensive checking
  • Double ring configuration
  • Constructed from durable tool steel
  • Comes with an inspection certificate traceable to NIST
  • Designed for right hand thread direction
  • Conforms to ASME B1.2 and ANSI standards
Catalog No. 50-005-36758 Supplier MSC Supplier No. 46553954
This threaded ring gage is designed for precise inspection of threads to ensure the proper assembly of mating components. It features an adjustable type for versatile use in various applications.
  • Adjustable design for flexible application
  • Ensures proper assembly of mating components through accurate thread inspection
  • Includes both Go and No Go functions for comprehensive checking
  • Double ring configuration
  • Constructed from durable tool steel
  • Comes with an inspection certificate traceable to NIST
  • Designed for right hand thread direction
  • Conforms to ASME B1.2 and ANSI standards