Learn More
MSC SPI Threaded Ring Gage, M3.5 x 0.60 Thread, Metric, Class 6G, No Go
Catalog No. 5000530165
Encompass
This threaded ring gage is designed for precise measurement of M3.5 x 0.60 metric threads. It is a No Go type gage, ensuring that components meet the specified Class 6G tolerance. Constructed from durable tool steel, it provides reliable performance and includes NPL traceability certification for quality assurance.
  • No Go functionality for accurate thread inspection.
  • M3.5 x 0.60 Metric thread for specific applications.
  • Class 6G classification for precise tolerance verification.
  • Constructed from tool steel for durability.
  • NPL traceability certification included for quality assurance.
Catalog No. 50-005-30165 Supplier MSC Supplier No. 75890293
This threaded ring gage is designed for precise measurement of M3.5 x 0.60 metric threads. It is a No Go type gage, ensuring that components meet the specified Class 6G tolerance. Constructed from durable tool steel, it provides reliable performance and includes NPL traceability certification for quality assurance.
  • No Go functionality for accurate thread inspection.
  • M3.5 x 0.60 Metric thread for specific applications.
  • Class 6G classification for precise tolerance verification.
  • Constructed from tool steel for durability.
  • NPL traceability certification included for quality assurance.