Learn More
MSC SPI Threaded Ring Gage, M5 x 0.80 Metric 6G, No Go
Catalog No. 5000528045
Encompass
This threaded ring gage is designed for precise measurement of M5 x 0.80 metric threads. It features a No Go design and is classified as 6G. Made from oil hardened non-shrinking tool steel, it ensures durability and accuracy, and includes NPL traceability certification.
  • No go functionality
  • M5 x 0.80 metric thread size
  • Class 6G classification
  • Single ring design
  • Made from tool steel
  • NPL traceability certification included
  • Country of origin: India
Catalog No. 50-005-28045 Supplier MSC Supplier No. 75890350
This threaded ring gage is designed for precise measurement of M5 x 0.80 metric threads. It features a No Go design and is classified as 6G. Made from oil hardened non-shrinking tool steel, it ensures durability and accuracy, and includes NPL traceability certification.
  • No go functionality
  • M5 x 0.80 metric thread size
  • Class 6G classification
  • Single ring design
  • Made from tool steel
  • NPL traceability certification included
  • Country of origin: India